Learn More
BrandTech™ VACUUBRAND™ Absolute Pressure Vacuum Gauges Available on GSA/VA Contract for Federal Government customers only.
Replace fragile McLeod and dial gauges
Manufacturer:  BrandTech™ 682902
 View more versions of this product
Catalog No. 13-878-81
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CE
General purpose lab applications
4.5 in.
9V Lithium battery with adjustable sleep timer, gauge, mounting rod, operating manual
6.3cm
Upper measuring limit:1080mbar / 810 torr; Lower measuring limit: 1 mbar / 1 torr
4.5 in.
Absolute Pressure Vacuum Gauge
2.5 x 4.5 x 4.5 in. (6.3 x 11.5 x 11.5cm)
Vacuum systems, diaphragm vacuum pumps and aspirators
11.5cm
2.5 in.
-10 to 40 deg.C
1mbar
11.5cm