Learn More
BrandTech™ VACUUBRAND™ BVC Fluid Aspiration Systems for Cell Culture Available on GSA/VA Contract for Federal Government customers only.
BVC fluid aspiration systems are the first to account for the needs of a modern biology laboratory
Manufacturer:  BrandTech™ 727400
 View more versions of this product
Catalog No. 13-688-011
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications
Description
  • with all the advantages of the BVC control
  • non-contact sensor for electronic monitoring of the liquid level in the collection bottle
  • disinfection routine for the suction tubing for drawing in disinfectant after switching off the pump
  • self-closing quick couplings (as accessory to the BVC professional G) for convenient and safe bottle change, e.g., for work with biohazards
  • for professional work and a perfect fit for required safety protocols
Description & Specifications
Specifications
0.4/0.5 cfm
IP40
45dBA
-10 to 60°C
7.3kg
50/60Hz
19.68 in.
7.63 in.
40.8cm
104°F
230V 50/60Hz
BVC professional
10° to 40°C
16.09 lb.
230V
14 to 140°F
50cm
16.06 in.
4L Polypropylene bottle, external liquid level sensor, CEE plug, 0.2μm biofilter
40°C