BrandTech™ VACUUBRAND™ CVC3000 detect Integrated Vacuum Control Systems Available on GSA/VA Contract for Federal Government customers only.
Vacuum controller with integrated VVB-6C solenoid valve for easy upgrading of vacuum systems to electronic control
Manufacturer: BrandTech™ 2614120
 View more versions of this product
Catalog No. 13-879-501
  Check Availability
Add to Cart
    Specifications
    Includes Stand, power supply, US, CEE, CH, UK and AU plugs and instructions for use, with C3 integrated solenoid valve
    Voltage 100/240V
    Hertz 50/60Hz
    Electrical Requirements 100/240V 50/60Hz
    Model Support Rod Version
  • Description & Specifications
Related Products
Product Content Feedback