Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
BrandTech™ VACUUBRAND™ NT Diaphragm Vacuum Pumps: ME4 NT Available on GSA/VA Contract for Federal Government customers only.
For a wide variety of non-corrosive and non-evaporative applications. ME-series pumps feature an ultimate vacuum of 52 Torr.
Manufacturer: BrandTech™ 731005
 View more versions of this product
Catalog No. 13-889-126
  N/A
Add to Cart
Related Products
Product Content Feedback