Learn More
BrandTech™ VACUUBRAND™ Rotary Vane Vacuum Pumps: Oil Mist Filters
Oil Mist Filters and replacements for VACUUBRAND Rotary Vane Vacuum Pumps
$132.00 - $1251.00
Catalog Number | Mfr. No. | For Use With | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With | Type | Price | Quantity & Availability | ||||
13-889-110
|
BrandTech™ VACUUBRAND™
20698003 |
Model RE2, RZ2, RE2.5, RZ2.5, RE5, RZ5, RE6, RZ6 rotary vane vacuum pump | Oil Mist Filter |
Each for $1,126.00
|
|
||||
13-889-111
|
BrandTech™ VACUUBRAND™
20698004 |
RZ 2.5 and RZ 6 Oil Mist Filter (20698003) | Filter Element for Oil Mist Filter |
Each for $132.00
|
|
||||
13-889-114
|
BrandTech™ VACUUBRAND™
20698017 |
Model RE8, RE9, RE16 and RZ8, RZ9, RZ16 rotary vane vacuum pump | Oil Mist Filter |
Each for $1,251.00
|
|
||||
Description
For use with VACUUBRAND Rotary Vane Vacuum Pumps
- Oil Mist Filters “FO” trap up to 99% of oil mist generated by the pump, helping preserve the laboratory environment
- Cat. No. 13-889-111 is a replacement part for small oil mist filter 13-889-110
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.