Learn More
BrandTech™ VITLAB™ PFA Evaporating Dishes Available on GSA/VA Contract for Federal Government customers only.
Highly resistant to heat and chemical contamination
Manufacturer:  BrandTech™ V103297
 View more versions of this product
Catalog No. 14-379-183
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.84 oz.
0.84 oz., 25mL
50mm
0.94 in.
PFA
-200° to 260°C
25mL
2 in.
For sterilization using heat, gas, and chemical methods for sensitive biological applications
24mm
-392° to 500°F
Evaporating Dish