Learn More
BrandTech™ VITLAB™ PFA Evaporating Dishes Available on GSA/VA Contract for Federal Government customers only.
Highly resistant to heat and chemical contamination
Manufacturer:  BrandTech™ V103397
 View more versions of this product
Catalog No. 14-379-184
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Evaporating Dish
50mL
2 in.
For sterilization using heat, gas, and chemical methods for sensitive biological applications
54mm
-392° to 500°F
1.69 oz.
1.69 oz., 50mL
50mm
2.12 in.
PFA
-200° to 260°C