Learn More
BrandTech™ VITLAB™ Polystyrene Graduated Cylinders Class B
Polystyrene graduated cylinders have ring molded graduations at major increments for excellent readability
Manufacturer:  BrandTech™ V64691
 View more versions of this product
Catalog No. 14-379-165
Add to cart
Description
Description
  • Styrene acrylonitrile copolymer (SAN)
  • Class B, per DIN 12681
  • Hexagonal base resists rolling
Specifications
Specifications
10mL
SAN (Styrene-acrylonitrile)
Plain molded graduations