Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Branson Ultrasonics Microtips for 200 and 400 Watt Sonifiers

Designed to process samples contained in small diameter vessels at extremely high intensity

Manufacturer:  Branson Ultrasonics 101148062

 View more versions of this product

Catalog No. 22-309796


  Check Availability
Add to cart

Alert:

Do not use in open air or at an output control setting greater than 7 on all Sonifier models as tips are easily breakable.


Specifications Description & Specifications

Specifications

116 to 494μm
0.3cm
Ultra high
Tapered
0.25 to 10mL
Tapered Microtip
0.12 in.
Ideal for processing spores, fungi, yeast, muscle and connective tissue
Titanium
0.25 to 10mL
200 and 400w Sonifier™ cell disruptor
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit