Learn More
Branson Ultrasonics™ SLP Cup Horn for Sonifiers SLPe, SLPt, 150E and 150T
3 in. diameter cup horn for the SLP Sonifier Cell Disruptor is ideal for processing single or multiple vials and test tubes with identical paramaters.
Manufacturer:  Branson Ultrasonics™ 1091161760
Catalog No. 15-335-130
Add to cart
Description
Description
Enables sonification of very small volumes without horn coming into contact with sample
  • Cup horn is attached to the inverted converter
  • Inlet and outlet enables liquid to circulate within the chip horn, to reduce heat buildup and overflow
  • Horn can also be used without liquid cooling circulation
  • Process samples in complete isolation
  • Ideal for infectious solutions
  • Precludes possibility of any titanium residue
  • Ideal for single or batch processing
  • Ideal for very small volume processing
Cell Disruption, Liposome preparation, Protein shearing and extraction, Releasing cellular components, including DNA and RNA, Tissue culture Infectious solutions
Specifications
Specifications
5 in.
Indirect method of disrupting
Titanium
Sonifier SLP, 150E, 150T
3 in.