Branson Ultrasonics Sonifier™ Cup Horns

For use with Branson Sonifier™ S-450 Cell Disruptors

Manufacturer: Branson Ultrasonics 101147048

 View more versions of this product

Catalog No. 22-309791

  Check Availability
Add to Cart

    Specifications

    Diameter (English) 3 in.
    Diameter (Metric) 7.6cm
    For Use With (Application) Ideal for liposome preparation, viruses and radioactive materials
    Material Titanium
    Size 3 in.
    For Use With (Equipment) Sonifier™ S-450 cell disruptor
    Type Cup Horn

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit