Learn More
Humboldt Bunsen Burner Brass Flame Spreaders
Manufacturer:  HUMBOLDT MANUFACTURING COMPANY H6640
 View more versions of this product
Catalog No. 03-995A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.43 in.
To produce a broad, flat, fan-shaped flame, in bending glass tubing and other similar applications
Brass
Standard
1.09cm
For bunsen burner
Burner Wing Top