Alfa Aesar™ Brass foil, alloy 260, 0.13mm (0.005 in.) thick, 15x45cm (5.9x18in)

Manufacturer: Alfa Aesar™ 13504KJ

Quantity 5 pieces

 View more versions of this product

Catalog No. AA13504KJ

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00171255
    Chemical Name or Material Brass foil, Alloy 260, 0.13mm (0.005 in.) thick, 15x45cm (5.9x18 in.)

    Specifications

    Quantity 5 pieces
    Odor Odorless
    TSCA TSCA
    Physical Form Foil
    MDL Number MFCD00171255
    Chemical Name or Material Brass foil, Alloy 260, 0.13mm (0.005 in.) thick, 15x45cm (5.9x18 in.)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit