Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Brass foil, alloy 260, 0.3mm (0.012 in.) thick

Manufacturer:  Alfa Aesar™ 45135HB

Quantity 300 x 300mm

 View more versions of this product

Catalog No. AA45135HB


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00171255
Brass foil, Alloy 260, 0.3mm (0.012 in.) thick

Specifications

300 x 300mm
Foil
Brass foil, Alloy 260, 0.3mm (0.012 in.) thick
Odorless
MFCD00171255

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit