Alfa Aesar™ Brass foil, alloy 260, 0.81mm (0.032 in.) thick

Manufacturer: Alfa Aesar™ 45138HB

Quantity 300 x 300mm

 View more versions of this product

Catalog No. AA45138HB

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00171255
    Chemical Name or Material Brass foil, Alloy 260, 0.81mm (0.032 in.) thick

    Specifications

    Quantity 300 x 300mm
    Odor Odorless
    TSCA TSCA
    MDL Number MFCD00171255
    Physical Form Foil
    Chemical Name or Material Brass foil, Alloy 260, 0.81mm (0.032 in.) thick

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit