Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCA1 Mouse anti-Human, Alexa Fluor™ 594, Clone: BRCA1/1472, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254337AF594

Catalog No. NP24337A94


Add to cart

Description

Description

BRCA1 Monoclonal antibody specifically detects BRCA1 in Human samples. It is validated for Flow Cytometry,ELISA,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

BRCA1
BRCA1/1472
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
BRCA1
Recombinant human BRCA1 protein fragment (aa445-620) (exact sequence is proprietary)
0.1 mL
Primary
672
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit