Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCA1 Mouse anti-Human, Clone: BRCA1/1472, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB15001

Catalog No. 89-035-021


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human BRCA1.

  • High specificity
  • High reactivity
Specifications

Specifications

BRCA1
BRCA1/1472
Mouse monoclonal antibody raised against partial recombinant human BRCA1.
In 10 mM PBS.
Liquid
BRCA1
Recombinant protein corresponding to amino acids 445-620 of human BRCA1.
Protein A/G purification
RUO
Primary
672
ELISA, Flow Cytometry, Immunofluorescence
Unconjugated
ELISA Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user.
P38398
BRCAI|BRCC1|IRIS|PSCP|RNF53
Mouse
IgG1
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)