Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRCA1, Mouse anti-Human, Clone: KEN, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280601
Description
BRCA1 Monoclonal antibody specifically detects BRCA1 in Human Samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation.Specifications
BRCA1 | |
KEN | |
Unconjugated | |
Tris-Glycine, 0.15 M NaCl | |
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
Mouse | |
IgG3 Kappa | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
Primary | |
672 | |
Human |
Immunofluorescence, Immunoprecipitation, Western Blot | |
1.67 mg/mL | |
Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500 | |
Purified | |
BRCA1 | |
Human BRCA1 corresponding to residues 1314-1864 [UniProt# P38398] | |
Protein A purified | |
RUO | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title