Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCA1 Mouse anti-Human, FITC, Clone: 6C6D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237361F

Catalog No. NBP237361F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BRCA1
6C6D2
ELISA, Immunohistochemistry
Purified
BRCA1
Purified recombinant fragment of human BRCA1 (AA: 229-335) expressed in E. Coli.
Protein A or G purified
Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger
Primary
672
ELISA, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit