Learn More
BRCA1 Mouse anti-Human, HRP, Clone: 6C6D2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237361H
Catalog No. NBP237361H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
BRCA1
6C6D2
Western Blot, ELISA, Immunohistochemistry
Purified
BRCA1
Purified recombinant fragment of human BRCA1 (AA: 229-335) expressed in E. Coli.
Protein A or G purified
Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger
Primary
672
ELISA, Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human