Learn More
BRCA2 Mouse anti-Human, Janelia Fluor 549, Clone: 5F6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141189JF549
Catalog No. NBP14189549
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
BRCA2Monoclonalantibody specifically detects BRCA2 in Human samples.
Specifications
Specifications
BRCA2
5F6
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
BRCA2
Amino acids 2336-2478 of human BRCA2 expressed in E. coli. BRCA2-P400.
Protein G purified
Breast Cancer, Cancer
Primary
675
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
BRCA1/BRCA2-containing complex, subunit 2, BRCC2, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast cancer susceptibility protein BRCA2, breast cancer type 2 susceptibility protein, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1Fanconi anemia, complementation group D1, Fanconi anemia group D1 protein, GLM3, PNCA2
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human BRCA2.