Learn More
BRCA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 197911AP
Catalog No. 50-173-2543
Add to cart
Description
Description
Inherited mutations in BRCA1 and this gene, BRCA2, confer increased lifetime risk of developing breast or ovarian cancer. Both BRCA1 and BRCA2 are involved in maintece of genome stability, specifically the homologous recombination pathway for double-strand DNA repair. The BRCA2 protein contains several copies of a 70 aa motif called the BRC motif, and these motifs mediate binding to the RAD51 recombinase which functions in DNA repair. BRCA2 is considered a tumor suppressor gene, as tumors with BRCA2 mutations generally exhibit loss of heterozygosity (LOH) of the wild-type allele.
Specifications
Specifications
BRCA2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35923, P51587, P97929
BRCA2, BRCC2, breast cancer 2, early onset, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1
Rabbit
IgG
150 μL
-20°C
Primary
12190, 360254, 675
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BRCA2
Liquid
Brca2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat