Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCAA1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132413
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA132413 100 μg
1 options

Catalog No. PIPA132413

Supplier: Thermo Scientific PA132413

Rabbit Polyclonal Antibody

PA1-32413 detects BRCAA1 from human samples. PA1-32413 has been successfully used in Western blot applications. The PA1-32413 immunogen is: Reacts with residues 1046-1055 [SSKKQKRSHK] of the 136 kDa human, mouse and rat BRCAA1 protein.

The novel BRCAA1 protein’s epitope, SSKKQKRSHK, was mapped to the cytoplasm of MCF-7 cells. Immunohistochemistry analysis showed that the antigen epitope exhibited positive expression in 65% of breast cancer specimens. Its expression is associated with
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCAA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene ARID4B
Gene Accession No. Q4LE39
Gene Alias Breast cancer-associated antigen BRCAA1,Rb-binding protein homolog,
Gene Symbols ARID4B
Host Species Rabbit
Immunogen Reacts with residues 1046-1055 [SSKKQKRSHK] of the 136 kDa human, mouse and rat BRCAA1 protein.
Purification Method IgG fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.