Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD 7389, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Ribosomal S6 kinase (RSK) inhibitor (IC50 values are 1.2, 1.5 and 2.4 μM for RSK3, RSK1 and RSK2 respectively). Upregulates insulin expression in terminally differentiated pancreatic α-cells.
Specifications
Quantity | 10 mg |
Formula Weight | 366.41 |
Percent Purity | >97% |
Chemical Name or Material | BRD 7389 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction