Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540885
Description
Peptide sequence: SGKKQAAKSK EELAQEKKKE LEKRLQDVSG QLSSSKKPAR KEKPGSAPSG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.
Specifications
BRD3 | |
Polyclonal | |
Unconjugated | |
BRD3 | |
2410084F24Rik; AW060456; BRD3; bromodomain containing 3; bromodomain containing 3; bromodomain-containing protein 3; bromodomain containing protein 3; bromodomain-containing 3; bromodomain-containing FSH-like protein FSRG2; Bromodomain-containing protein 3; bromodomain-containing protein 3 short isoform; Fsrg2; KIAA0043; ORFX; RING3L; RING3-like protein; RINGL3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8019 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15059 | |
BRD3 | |
Synthetic peptide directed towards the middle region of human BRD3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction