Learn More
BRD4 Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, Active Motif™
Rabbit Polyclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 39010
Encompass
 View more versions of this product
Catalog No. 50-199-2951
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
Specifications
Specifications
BRD4
Lot-Specific
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
O60885, Q9ESU6
BOS_7204; Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; Bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; Mitotic chromosome-associated protein; Protein HUNK1; WI-11513
This BRD4 antibody was raised against a peptide within the C-terminal region of human BRD4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ChIP assay, Immunohistochemistry, Western Blot
Unconjugated
BRD4
Liquid
Rabbit
IgG
50 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
23476, 57261
BRD4