Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD4 Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, Active Motif™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$135.30 - $546.00

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen BRD4
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2950 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39910
10 μL
Each for $135.30
Add to cart
 
50-199-2951 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39010
50 μL
Each for $320.65
Add to cart
 
50-199-2949 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39909
100 μL
Each for $546.00
Add to cart
 
Description

Description

BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
Specifications

Specifications

Polyclonal
Primary
BRD4
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit