Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRD7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549379
Description
BRD7 Polyclonal Antibody for Western Blot
BRD7 acts both as coactivator and as corepressor. For example, it activates the Wnt signaling pathway in a DVL1-dependent manner by negatively regulating the GSK3B phosphotransferase activity. In addition, it is required for TP53-mediated cell-cycle arrest in response to oncogene activation. BRD7 promotes acetylation of TP53 at Lys-382, and thereby promotes efficient recruitment of TP53 to target promoters, inhibiting cell cycle progression from G1 to S phase.
Specifications
BRD7 | |
Polyclonal | |
Unconjugated | |
BRD7 | |
75 kDa bromodomain protein; BP75; BRD7; bromodomain containing 7; bromodomain protein 75 kDa; bromodomain-containing 7; bromodomain-containing 7-like protein; bromodomain-containing protein 7; CELTIX1; NAG4; protein CELTIX-1; protein tyrosine phosphatase, non-receptor type 13 interacting protein; Ptpn13ip | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
26992 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O88665 | |
BRD7 | |
KLH conjugated synthetic peptide between 387-418 amino acids from the Central region of human MOUSE Brd7. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction