Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRD9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582820
Description
Immunogen sequence: DDSHLNLDET TKLLQDLHEA QAERGGSRPS SNLSSLSNAS ERDQHHLGSP SRLSVGEQPD VTHDPYEFLQ S.
Bromodomain containing protein 9 (BRD9) is an uncharacterized protein that contains one bromodomain.
Specifications
BRD9 | |
Polyclonal | |
Unconjugated | |
BRD9 | |
AL022779; Brd9; bromodomain containing 9; bromodomain-containing protein 9; DKFZp434D0711; DKFZp686L0539; FLJ43530; LAVS3040; PRO9856; rhabdomyosarcoma antigen MU-RMS-40.8; sarcoma antigen NY-SAR-29; UNQ3040/PRO9856 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
65980 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H8M2 | |
BRD9 | |
Recombinant protein corresponding to Human BRD9. Recombinant protein control fragment (Product #RP-105753). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction