Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRDU Chicken, Polyclonal , Abnova™

Chicken polyclonal antibody raised against BrdU.

Manufacturer:  Abnova Corporation PAB29679

Catalog No. 89-933-667


Add to cart

Description

Description

Specifications

Specifications

BRDU
Unconjugated
ELISA Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
BrdU
Antigen Affinity Purification
RUO
Primary
ELISA, Immunohistochemistry
Chicken polyclonal antibody raised against BrdU.
In PBS, pH 7.2 (0.09% sodium azide)
Chicken
IgY
100 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit