Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrdU Monoclonal Antibody (Bu20a), Active Motif™
Mouse Monoclonal Antibody
$147.50 - $565.00
Specifications
Antigen | BrdU |
---|---|
Clone | Bu20a |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3292
|
Active Motif
61274 |
10 μg |
Each for $147.50
|
|
|||||
50-199-3291
|
Active Motif
61273 |
100 μg |
Each for $565.00
|
|
|||||
Description
Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.Specifications
BrdU | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
2432 | |
This BrdU antibody was raised against Bromodeoxyuridine conjugated to BSA and recognizes BrdU. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
FRA10B |
Bu20a | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Multi-species, Mouse, Rat | |
5-Bromo-2-deoxyuridine; Bromodeoxyuridine | |
FRA10B | |
IgG1 | |
Protein A | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title