Learn More
BrdU Mouse anti-Chemical, Clone: OTI2G6, lyophilized, TrueMAB

Mouse Monoclonal Antibody
Supplier: OriGene CF190133

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA.Specifications
BrdU | |
Monoclonal | |
Unconjugated | |
BrdU; 5-Bromo-2-deoxyuridine; Bromodeoxyuridine | |
Bromodeoxyuridine coupled to keyhole limpet hemocyanin (KLH). | |
100 μg | |
Primary | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI2G6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized | |
Chemical |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.