Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrdU Mouse anti-Chemical, Clone: OTI7H4, liquid, TrueMAB

Mouse Monoclonal Antibody
Supplier: OriGene TA190207

Description
Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA.Specifications
BrdU | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Mouse | |
Affinity Chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI7H4 | |
Unconjugated | |
BrdU; 5-Bromo-2-deoxyuridine; Bromodeoxyuridine | |
Bromodeoxyuridine coupled to keyhole limpet hemocyanin (KLH). | |
100 μL | |
Primary | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction