Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Breast carcinoma amplified sequence 3 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298807PE
Description
Breast carcinoma amplified sequence 3 Polyclonal antibody specifically detects Breast carcinoma amplified sequence 3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Breast carcinoma amplified sequence 3 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
BCAS4/BCAS3 fusion, breast carcinoma amplified sequence 3, breast carcinoma amplified sequence 4/3 fusion protein, breast carcinoma-amplified sequence 3, DKFZp686O1527, FLJ20128, GAOB1, MAAB, metastasis associated antigen of breast cancer, MGC4973, protein Maab1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Breast carcinoma amplified sequence 3. | |
0.1 mL | |
Cancer | |
54828 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction