Learn More
3M™ Breathe Easy™ 12 Bio Conversion Kit (Regular)
Ideal for converting your current Breathe Easy™ Powered Air Purifying Respirator system that uses the BE-10BR Butyl Rubber Hood and the FR-57 cartridge for chemical protection
Manufacturer:  3M™ 7010384032
Catalog No. 19-070-836
Add to cart
Description
Description
  • BE-12-3 Head Covers are white in regular size
  • Set of HEPA filters for protection against biological hazards
  • 3M Catalog# 70071517026
Specifications
Specifications
Safety
Hood
Three head covers, a breathing tube, and a set of HEPA filters
1
Breathe Easy™ Powered Air Purifying Respirator systems