Learn More
American Educational Products Breathing Fitness Lab
SCI_ED
How much CO2 do you exhale?
Specifications
For Use With (Application) Investigate the pulmonary system by measuring breath rate, volume, recovery, control, lung volume and the amount of co2 in exhaled air
For Use With (Equipment) Breathing fitness
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
S16200
SCI_ED
View Documents Promotion Details
American Educational Products
6212
Breathing Fitness Health Lab Breath control meter, lung volume bag, breath volume bag and valve, meter tape, mouth pieces, chemicals, 3 charts, teacher packet with activity plans This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Investigate the pulmonary system by measuring breath rate, volume, recovery, control, lung volume and the amount of CO2 in exhaled air
  • Kit includes necessary equipment and teacher and student guides
Specifications
Specifications
Investigate the pulmonary system by measuring breath rate, volume, recovery, control, lung volume and the amount of co2 in exhaled air
Breathing fitness