Learn More
Sklar InstrumentsSupplier Diversity Partner Breisky Retractor, Sklar, 13 in. Breisky Retractor 130mm X 40mm, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 903528
Breisky Retractor, Sklar, 13 in. Breisky Retractor 130mm X 40mm, Stainless Steel. Sklar's Breisky Retractor is a bayonet shaped instrument commonly used to aid in the examination of the vaginal walls. The concave blade is curved back from the grip handle to provide a larger opening for the exam. The retractor is available in a variety of lengths and blade sizes to accommodate the needs for a wide range of procedures.
Catalog No. 50-143-6640