Learn More
Brentuximab, Rabbit anti-Many, Alexa Fluor 700, Clone: 2262C, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB9584N100UG
Catalog No. FAB9584N100
Add to cart
This item is not returnable. View return policy
Description
Description
Brentuximab Monoclonal antibody specifically detects Brentuximab in Multi-Species Samples. It is validated for Flow Cytometry.
Specifications
Specifications
Brentuximab
2262C
Flow Cytometry 0.25-1 ug/10^6 cells
Purified
Full length Brentuximab
Protein A or G purified from cell culture supernatant
RUO
Primary
Detects Brentuximab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Trastuzumab, Tositumomab, Adalimumab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed.
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Rabbit
IgG
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Many