Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Brentuximab (Anti-Idiotype) Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB9584S100UG
This item is not returnable.
View return policy
Description
Brentuximab Monoclonal specifically detects Brentuximab in Multi-Species samples. It is validated for Flow Cytometry.Specifications
Brentuximab | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Full length Brentuximab | |
100 μg | |
Primary | |
Multi-species | |
Purified |
Flow Cytometry | |
2262C | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Detects Brentuximab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Trastuzumab, Tositumomab, Adalimumab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction