Learn More
Aviva Systems Biology Corp BRF2 Antibody - N-terminal region (ARP50630_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50630_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BRF2. Alternate names: BRFU, FLJ11052, TFIIIB50ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60392