Learn More
BRG1 Rabbit anti-Human,Mouse, Unconjugated, Clone: 1N12, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB114825UL
 View more versions of this product
Catalog No. ZRB114825MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
BRG1 Monoclonal antibody specifically detects BRG1 in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
BRG1
1N12
Unconjugated
Purified
SMARCA4, BAF190A, BRG1, SNF2B, SNF2L4
His-tagged recombinant fragment corresponding to 65 amino acids from the N-terminal region of human Transcription activator BRG1.
Target molecular weight 184.65 kDa calculated; ∼185 kDa observed. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_001122316.1
Transcription activator BRG1;ATP-dependent helicase;SMARCA4;BRG1-associated factor 190A;BAF190A;Mitotic growth and transcription activator;Protein BRG-1;Protein brahma homolog 1;SNF2-beta;SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4
Rabbit
IgG
Protein A Purified
Epigenetics and Nuclear Function
Primary
Human, Mouse