Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRG1 Rabbit anti-Human,Mouse, Unconjugated, Clone: 1N12, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen BRG1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB114825MI
View Documents
MilliporeSigma
ZRB114825UL
25 μL N/A N/A N/A
ZRB11484XMI
View Documents
MilliporeSigma
ZRB11484X25UL
4 x 25 μL N/A N/A N/A
Description

Description

BRG1 Monoclonal antibody specifically detects BRG1 in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

BRG1
Unconjugated
Rabbit
Transcription activator BRG1;ATP-dependent helicase;SMARCA4;BRG1-associated factor 190A;BAF190A;Mitotic growth and transcription activator;Protein BRG-1;Protein brahma homolog 1;SNF2-beta;SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4
His-tagged recombinant fragment corresponding to 65 amino acids from the N-terminal region of human Transcription activator BRG1.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse
SMARCA4, BAF190A, BRG1, SNF2B, SNF2L4
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit