Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRI3BP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553261
Description
Immunogen sequence: MFVETLWKVW TELLDVLGLD VSNLSQYFSP ASVSSSPARA LLLVGVV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.
BRI3BP (BRI3 Binding Protein) is a Protein Coding gene located in mitochondrion. Involved in tumorigenesis and may function by stabilizing p53/TP53.
Specifications
BRI3BP | |
Polyclonal | |
Unconjugated | |
BRI3BP | |
2410150I18Rik; AI841257; AW742481; BNAS1; BRI3 binding protein; BRI3-binding protein; Bri3bp; cervical cancer 1 proto-oncogene-binding protein KG19; cervical cancer oncogene binding protein; HCCR-1; HCCR-2; HCCRBP-1; I3-binding protein; KG19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140707 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WY22 | |
BRI3BP | |
Recombinant protein corresponding to Human BRI3BP. Recombinant protein control fragment (Product #RP-90545). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction