Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads - Promo

Color-coded absorbents for safe clean-up of hazardous fluids. Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads are surfactant treated polypropylene pads used on wide range of chemicals.

Manufacturer:  Brady™ CH200PM

 View more versions of this product

Catalog No. 19-103-101PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Benefits of sign and an absorbent in one
  • Highly absorbent for all-purpose industrial applications
  • Ideal for areas where the risk of slips, trips and falls are greatest
  • Chemically inert, do not react with aggressive fluids
  • Perforations provide cost control and reduce waste
  • Heavy weight - specifically designed for continuous leaks and larger spills
  • Light weight - specifically designed for smaller leaks, drips and spills
Specifications

Specifications

Polypropylene
38cm
Perforated
Black/Red on Yellow
106L
19 in.
Light
6.8kg
19 x 15 in. (48.2 x 38cm)
15 in.
Does Not Contain Recycled Content
EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
28 gal.
48.2cm
Absorbent Pad
15 lb.
28 gal., 106L
200/Cs.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit