Learn More
Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads - Promo
Color-coded absorbents for safe clean-up of hazardous fluids. Brady™ BRIGHTSORB™ High Visibility Chemical Absorbent Pads are surfactant treated polypropylene pads used on wide range of chemicals.
Specifications
Certifications/Compliance EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
Color Black/Red on Yellow
Dimensions (L x W) 19 x 15 in. (48.2 x 38cm)
Item Description Does Not Contain Recycled Content
Material Polypropylene
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-103-101PM
View Documents
Brady™
CH200PM
N/A Light-weight; Absorbancy: 28 gal.; 200/Cs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Benefits of sign and an absorbent in one
  • Highly absorbent for all-purpose industrial applications
  • Ideal for areas where the risk of slips, trips and falls are greatest
  • Chemically inert, do not react with aggressive fluids
  • Perforations provide cost control and reduce waste
  • Heavy weight - specifically designed for continuous leaks and larger spills
  • Light weight - specifically designed for smaller leaks, drips and spills
Specifications
Specifications
EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
19 x 15 in. (48.2 x 38cm)
Polypropylene
6.8kg
38cm
48.2cm
Perforated
Black/Red on Yellow
Does Not Contain Recycled Content
15 lb.
15 in.
19 in.
Absorbent Pad