Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij(R) S100, Spectrum™

Click to view available options
Quantity:
25 g
500 g
2.5 kg
30 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
9005-00-9
C20H41O2
313.55
PSEGVHKUPXNGGJ-UHFFFAOYSA-N
[2-(octadecyloxy)ethyl]oxidanyl
CCCCCCCCCCCCCCCCCCOCC[O]
9005-00-9
C20H41O2
313.55
PSEGVHKUPXNGGJ-UHFFFAOYSA-N
[2-(octadecyloxy)ethyl]oxidanyl
CCCCCCCCCCCCCCCCCCOCC[O]

B1593, 9005-00-9

Spectrum™ Brij(R) S100 , Polyethylene (100) Stearyl Ether, functions as an aqueous dispersant and oil/water emulsifier.
CAS Max % 1
Packaging Fiber Drum
Quantity 30 kg