Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brij(R) S2, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Pail
Poly-Coated Glass Bottle
9005-00-9
C20H41O2
313.55
PSEGVHKUPXNGGJ-UHFFFAOYSA-N
[2-(octadecyloxy)ethyl]oxidanyl
CCCCCCCCCCCCCCCCCCOCC[O]
9005-00-9
C20H41O2
313.55
PSEGVHKUPXNGGJ-UHFFFAOYSA-N
[2-(octadecyloxy)ethyl]oxidanyl
CCCCCCCCCCCCCCCCCCOCC[O]

B1670, 9005-00-9

Spectrum™ Brij(R) S2 or Polyoxyethylene (2) Stearyl Ether are designed to emulsify and produce stable dispersions of cosmetic materials. It appears as a colorless to light yellow liquid that may become hazy or form a precipitate.
CAS Max % 100%
Appearance White waxy solid
Packaging Poly Pail
Quantity 12 kg