Learn More
Brij(R) S2, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Drum
Poly Pail
Poly-Coated Glass Bottle
B1670, 9005-00-9
Spectrum™ Brij(R) S2 or Polyoxyethylene (2) Stearyl Ether are designed to emulsify and produce stable dispersions of cosmetic materials. It appears as a colorless to light yellow liquid that may become hazy or form a precipitate.
Packaging Drum