Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brij(R) S2, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
Poly-Coated Glass Bottle
Description
Spectrum™ Chemical Brij(R) S2 or Polyoxyethylene (2) Stearyl Ether are designed to emulsify and produce stable dispersions of cosmetic materials. It appears as a colorless to light yellow liquid that may become hazy or form a precipitate.
Specifications
Color | White waxy solid |
CAS Max % | 100% |
Quantity | 125 g |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction