Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Blue G, Concentrate, Spectrum™

Click to view available options
Quantity: 1/Ea
CAS 6104-58-1
CAS Max % 1
Grade Chemical Solution
Quantity 1 ea

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.